Learn More
Thermo Scientific™ Validation Tools for UV-Visible Spectrophotometers
Product Code. 16665492 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
GENESYS Validator
Performance Verification Kit
Validator 2 for INSIGHT - A4
Validator Lite
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
16665492 Performance Verification Kit Each
11947471 Validator Lite Each
15905352 Validator 2 for INSIGHT - A4 Each
16594560 GENESYS Validator Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16665492 Supplier Thermo Scientific™ Supplier No. 840326100
  • ONLINE EXCLUSIVE PRICE
    2465.00€
    2302.65€ / pezzi 1
    Risparmia 162.35€
    6% Sconto
Spedizione stimata: 21-01-2027
Ensure compliance with regulatory guidelines with the Thermo Scientific™ Validation Tools for UV-Visible Spectrophotometers providing tools and documentation for system qualification.
Get all of the documentation and reference materials needed to facilitate Installation and Operational qualification procedures (IQ/OQ) with Thermo Scientific™ Validation Tools for UV-Visible Spectrophotometers. Certified standards for verifying instrument performance to factory specifications are available separately.
UV-Visible Spectrophotometer Validation Solutions include:
  • Information on the validation process
  • Documentation for manufacturer-verified Installation Qualification (IQ) to ensure proper system performance
  • Information on pharmacopoeia recommended tests for specific instrument performance where applicable
  • Installation and instrument certification
Solutions available for GENESYS 40-180, BioMate 160, Evolution200 series, Evolution 350, NanoDrop One, NanoDrop 2000, and NanoDrop 8000 instruments
TRUSTED_SUSTAINABILITY
Specifications
Type Performance Verification Kit
For Use With (Equipment) GENESYS 40-180, BioMate 160 Spectrophotometers