Learn More
Valproic Acid, TRC
Codice prodotto. 30041032
Cambia vista
Click to view available options
Quantità:
100 mg
1 g
10 g
Dimensione della confezione:
100mg
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30041032 10 g 10g
30040917 1 g 1g
30040886 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Codice prodotto. 30041032 Fornitore Toronto Research Chemicals N. del fornitore TRCV09475010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 99-66-1
Formula molecolare C8 H16 O2
Molecular Weight (g/mol) 144.21
Sinonimo Valproic Acid, 2-Propylpentanoic acid, 2-Propylvaleric acid, 4-Heptanecarboxylic acid, 44089, Acetic acid, dipropyl-, DPA, Depakine, Dipropylacetic acid, Ergenyl, Mylproin, NSC 93819, Valproic acid, n-Dipropylacetic acid
IUPAC Name 2-propylpentanoic acid
SMILES CCCC(CCC)C(=O)O
Specifica
Materiale o nome chimico Valproic acid
Note sul grado di purezza GC
Quantità 10 g
InChI Formula InChI=1S/C8H16O2/c1-3-5-7(6-4-2)8(9)10/h7H,3-6H2,1-2H3,(H,9,10)
Peso formulazione 144.115
Percent Purity >95%