Learn More
Brady™ Valve and Electrical Lockout Kit
Product Code. 15613547 Shop All Brady Products
Change view
Click to view available options
Color:
Red/Silver
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
15613547 Red/Silver Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15613547 Supplier Brady™ Supplier No. 806179
Spedizione stimata: 03-12-2026
The combination lockout kit gives you everything you need to lock out the most common mechanical and electrical control points.
  • Kit for valve and electrical lockout
Specifications
Alternate Part Number Valve and Electrical Lockout Kit (NL)
Color Red/Silver
Includes Large Universal Valve Lockout base clamping unit, Large blocking arm, Cable attachment, Adjustable gate valve device, APLCO with sheathed metal cable, Large plug lockout device, 227 V No-hole circuit breaker, 480 - 600 V No-hole circuit breaker, 480 - 600 V Breaker blocker kit, Miniature circuit breaker demo kit (1 off each type), Nylon lockout hasp, 1 Pk. of red safety padlocks, 1 Pk. of red safety lockouts (25 mm), Do not operate tag and pen. All supplied in a 600 denier polyester duffel bag.