missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VAMP4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10738-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
The vesicle associated membrane proteins (VAMP) or synaptobrevins are calcium binding proteins specific to eukaryotes. VAMPs, along with synaptosomal associated protein of 25 kDa (SNAP-25) and syntaxin, form the core complex of soluble NSF attachment protein receptor (SNARE) proteins that interact with the soluble proteins N-ethylmaleimide-sensitive factor (NSF) and alpha-SNAP. These membrane associated proteins play a key role in the regulation of vesicle membrane fusion with the plasma membrane. The Clostridium tetani neurotoxin is a metalloprotease with specificity for VAMP. In Alzheimer′s disease, VAMP levels of all isoforms appear to be significantly lowered.Specifica
VAMP4 | |
Polyclonal | |
Unconjugated | |
VAMP4 | |
VAMP 4, VAMP24, VAMP4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
364033, 53330, 8674 | |
-20°C | |
Liquid |
Western Blot, Immunofluorescence, Immunoprecipitation, Immunocytochemistry | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O70480, O75379 | |
Vamp4 | |
VAMP4 Fusion Protein Ag1090 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |