Learn More
Macherey-Nagel™ Supporto per precolonna VarioPrep
Codice prodotto. 12757690 Sfoglia Tutto Macherey-Nagel Prodotti
Cambia vista
Click to view available options
Diametro (sistema metrico) interno:
8 mm
32 mm
Lunghezza (sistema metrico):
10 mm
15 mm
Dimensione della confezione:
Pezzo
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Diametro (sistema metrico) interno Lunghezza (sistema metrico) Dimensione della confezione
12757690 32 mm 15 mm Pezzo
12472610 8 mm 10 mm Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 12757690 Fornitore Macherey-Nagel™ N. del fornitore 718253
Spedizione stimata: 10-12-2026
Use VarioPrep™ stainless steel columns for HPLC. VarioPrep™ Guard Column Holder for Preperative HPLC come in different sizes for a variety of columns.
Features
  • Column system for preparative HPLC manufactured from stainless steel with two adjustable end fittings, e.g., for frequent use of backflushing techniques
  • Allows compensation of a dead volume, which could result at the column inlet after some time of operation, without need for opening the column
  • Can be packed with all NUCLEODUR™ and NUCLEOSIL™ spherical silicas
  • Column holders come in a variety of sizes for different columns
Specifica
Diametro (sistema metrico) interno 32 mm
Lunghezza (sistema metrico) 15 mm
Product Type Column Holder