missing translation for 'onlineSavingsMsg'
Learn More

VCY Antibody, Novus Biologicals™

Codice prodotto. 18694516 Sfoglia Tutto Bio Techne Prodotti
Cambia vista
Click to view available options
Quantità:
0.1 mL
25 μL
Dimensione della confezione:
0.10mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
18694516 25 μL 25µL
18676565 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 18694516 Fornitore Novus Biologicals N. del fornitore NBP24670725ul

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

VCY Polyclonal antibody specifically detects VCY in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigene VCY
Applicazioni Immunohistochemistry, Immunohistochemistry (Paraffin)
Classificazione Polyclonal
Coniugato Unconjugated
Diluizione Immunohistochemistry 1:2500 - 1:5000, Immunohistochemistry-Paraffin 1:2500 - 1:5000
Formulazione PBS (pH 7.2), 40% Glycerol
Alias gene Basic Charge, Y-Linked 1, Basic Protein On Y Chromosome, 1, BPY1, BPY1B, Testis-Specific Basic Protein On Y, 1, Testis-Specific Basic Protein Y 1, Variable Charge, Y Chromosome, Variable Charge, Y-Linked, Variably Charged Protein Y, VCY VCY1B, VCY1, VCY1A
Specie ospite Rabbit
Immunogeno This antibody was developed against Recombinant Protein corresponding to amino acids: AATKMAAVTAPEAESGPAAPGPSDQPSQE
Metodo di purificazione Immunogen affinity purified
Quantità 25 μL
Status giuridico RUO
Primario o secondario Primary
ID gene (immissione) 9084
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Forma Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.