Learn More
3M™ Versaflo™ Intrinsically Safe Powered Air Turbo TR-802E
Product Code. 16416370 Shop All 3M Products
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16416370 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16416370 Supplier 3M™ Supplier No. 7100169179
  • ONLINE EXCLUSIVE PRICE
    1180.00€
    919.40€ / pezzi 1
    Risparmia 260.60€
    22% Sconto
 Restricted Item
Spedizione stimata: 11-02-2027
3M™ Versaflo™ Intrinsically Safe Powered Air Turbo TR-802E is approved to Zone 0 requirements, making it suitable for areas where potentially explosive atmospheres are expected in normal operations.
  • Approved against the latest Electrical Standards
  • Easy to identify and differentiate from non-IS versions thanks to bright yellow touchpoints
  • Comfort is maximised with three user selectable flow rates and controlled airflow
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Ea.