Learn More
behr Labor-Technik™ Vessels
Product Code. 10516752
Change view
Click to view available options
Capacity (Metric):
250 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10516752 250 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10516752 Supplier behr Labor-Technik™ Supplier No. B00217947
  • ONLINE EXCLUSIVE PRICE
    117.42€
    74.81€ / pezzi 1
    Risparmia 42.61€
    36% Sconto
 Restricted Item
Spedizione stimata: 09-12-2026
Use these vessels with behrotest™ SMA workstations. behr Labor-Technik™ Vessels are ideal for extraction applications.
  • SR 3 digestion vessels
  • Wide opening
  • Reinforced walls in the lower area
  • PVDF collars
  • 42mm dia.
  • Round bottom
Order Info
CapacityMetric
Specifications
Product Type Vessel
Capacity (Metric) 250 mL
For Use With (Equipment) behrotest™ SMA workstations
Material PVDF collar