Learn More
Veolia Sievers Certified TOC Vials
Product Code. 13287930
Change view
Click to view available options
Color:
Clear
Unit Size:
Pack of 72
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
13287930 Clear Pack of 72
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13287930 Supplier Veolia Supplier No. CHMI90606
Top Sellers. Mega Savings.
Use promo code 27997
View all promotions
In stock
Add to basket
Edge
Add to basket
Use these certified TOC vials in critical pharmaceutical applications such as USP and EP Water testing as well as municipal water testing. Veolia Sievers Certified TOC Vials are certified to below 10 ppb for confidence in the quality of your TOC measurements.
  • Sievers Certified (<10 ppb TOC) Vials
  • 72 x 40mL glass vials with caps and Teflon-lined septa, 72 per case
  • Ideal for strict regulatory compliance testing
  • Scrupulously cleaned in an ISO 9001 environment using a validated, automated process
  • Manufactured by the global leader in TOC measurement technology and TOC standards
  • Economical, with added savings on volume orders
  • Reliable, with guaranteed performance
  • Compatible with most TOC analyzer brands, including Sievers, Shimadzu, OI Analytical, Teledyne-Tekmar and Skalar
Specifications
Color Clear
Material Glass
Volume (Metric) 40 mL
For Use With (Application) Certified for use in critical applications including USP, EP, JP, and KP water testing
No. per Case 72
Certifications/Compliance TOC Certified (<10 ppb)