Learn More
Brady™ Vinyl with Polyester Overlaminate Stripes
Marca: Brady™ 090019
Dettagli aggiuntivi : Peso : 0.20410kg
Specifica
1 Roll | |
Rubber-based Pressure Sensitive Adhesive | |
Safety marking of obstructions, low hanging objects, pillars, passageways, aisles, and any other potentially hazardous areas or objects | |
50.8 mm | |
-40°C to +80°C |
0.037 mm | |
Green, White | |
Stripes | |
Vinyl | |
16.45 m |