Learn More
Thermo Scientific™ VISION pro™ UV-Visible Spectrophotometer VISIONpro Software, Single license
Thermo Scientific™ VISION pro™ is full-featured instrument control software for applications from research to routine analysis.
Marca: Thermo Scientific™ 10040101
Dettagli aggiuntivi : Peso : 50.00000kg
Descrizione
Thermo Scientific™ VISION pro™ is full-featured instrument control software for applications from research to routine analysis. Powerful, versatile software offers advanced instrument control and data manipulation capabilities. Optimized for Thermo Scientific UV-Visible spectrophotometers, it brings a user-friendly, graphical environment to spectrophotometric analysis. Method setup, data collection, calculations, report presentation, regulatory compliance - all aspects of your analysis are easier with this software.
- For use with Thermo Scientific Evolution™ 100, 160, 300 and 600 UV-Vis Spectrophotometers
- Performs Scan, Fixed Wavelength Analysis, Quantification and Multi-Component Analysis
- Produces spreadsheet results table
- Arranges data easily with report composer
Includes:
User Manual and RS-232-C cable (Part No. 401317263061)
Requires:
System Requirements: Microsoft™ Windows™ 2000, XP, Vista
Specifica
FDA guidelines, IEEE software engineering standards | |
VISIONpro Software; Single License | |
Fixed, Scan, Quant and MCA |
10040101 | |
Evolution 300 | |
At least Windows XP |