Learn More
VOC Mixture 561 1000 μg/mL in Methanol, Dr. Ehrenstorfer
Codice prodotto. 30328008
Cambia vista
Click to view available options
Quantità:
1 mL
Dimensione della confezione:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30328008 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 30328008 Fornitore Dr. Ehrenstorfer N. del fornitore DREA50000561ME
Please to purchase this item. Need a web account? Register with us today!
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifica
CAS 71-43-2, 123-86-4, 100-41-4, 108-38-3, 544-76-3, 110-54-3
Quantità 1 mL
Certificazione/Conformità ISO 17034
Solvente o matrice Methanol