Learn More
VPREB3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA551806
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: QLDALLVFPG QVAQLSCTLS PQHVTIRDYG VSWYQQRAGS APRYLLYYRS EEDHHRPADI PDRFSAAKDE AHNACVLTIS PVQPEDDADY YCSVGYGFSP Highest antigen sequence identity to the following orthologs: Mouse - 69%, Rat - 58%.
VPREB3 Polyclonal antibody specifically detects VPREB3 in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifica
VPREB3 | |
Polyclonal | |
Unconjugated | |
VPREB3 | |
8HS20; N27C7-2; pre-B lymphocyte 3; pre-B lymphocyte gene 3; pre-B lymphocyte protein 3; Protein VPreB3; UNQ355/PRO619; VPREB3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
29802 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UKI3 | |
VPREB3 | |
Recombinant protein corresponding to Human VPREB3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only