Learn More
Haleco™ Waste Disposal Bags
Product Code. 12526239
Change view
Click to view available options
Capacity (Metric):
50 L
100 L
Unit Size:
Pack of 200
Pack of 500
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
12526239 50 L Pack of 500
12536239 100 L Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12526239 Supplier Haleco™ Supplier No. SACG50ST
  • ONLINE EXCLUSIVE PRICE
    92.90€
    79.89€ / pezzi 500
    Risparmia 13.01€
    14% Sconto
View all promotions
 Restricted Item
Spedizione stimata: 15-12-2026
Specifications
Color Gray
Disposable Single-use
For Use With (Application) Multi-purpose
Material LDPE
Weight (Metric) 2.5 kg
Product Type Garbage Bag
Capacity (Metric) 50 L
Length (Metric) 870 mm
Width (Metric) 600 mm
Thickness (Metric) 140 μm