Learn More
Water, LC-MS Grade
Product Code. 18871496
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18871496 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18871496 Supplier Thermo Scientific Alfa Aesar Supplier No. 047146.K2
 Restricted Item
In stock
Add to basket
Edge
Add to basket
Mobile phase solvent in LC-MS analysis | CAS: 7732-18-5 | H2O | 18.015 g/mol
It is used as mobile phase solvent in LC-MS analysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
MDL Number MFCD00011332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O
Specifications
Color Colorless
Melting Point 0°C
Boiling Point 100°C
Beilstein 2050024
Merck Index 14,10039
Solubility Information Soluble in water. Insoluble in polar liquids.
Formula Weight 18.02
Grade LC-MS
Odor Odorless
Density 1.000 g/mL
Quantity 1 L
Refractive Index 1.34
Physical Form Liquid
Chemical Name or Material Water