Learn More
Thermo Scientific™ Water Purification Systems Storage Reservoirs
Codice prodotto. 13498709 Sfoglia Tutto Thermo Scientific Prodotti
Cambia vista
Click to view available options
Tipo:
Capacità 60l
Feed Water Tank
Sterile Overflow
Storage Reservoir
Capacità (sistema metrico):
100 L
30 l
30 to 100 L
30 l
60 L
60 l
Include:
Level Display
Level Display, Recirculation Pump
N/A
Reservoir
Tank
Da utilizzare con (apparecchiatura):
GenPure, GenPure Pro, GenPure xCAD
Pacific RO, Pacific TII
Pacific TII
Smart2Pure
Smart2Pure 12
Smart2Pure, Pacific and LabTower Water Purification Systems
Dimensione della confezione:
Pezzo
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Tipo Capacità (sistema metrico) Include Da utilizzare con (apparecchiatura) Dimensione della confezione
13498709 Sterile Overflow 30 to 100 L N/A Smart2Pure Pezzo
11377081 Storage Reservoir 30 l Level Display Pacific RO, Pacific TII Pezzo
11367061 Sterile Overflow 30 to 100 L N/A Smart2Pure, Pacific and LabTower Water Purification Systems Pezzo
11327091 Storage Reservoir 60 L Level Display, Recirculation Pump Pacific TII Pezzo
11367081 Storage Reservoir 30 l Level Display, Recirculation Pump Pacific TII Pezzo
15861448 Capacità 60l 60 l Reservoir Smart2Pure 12 Pezzo
11317091 Storage Reservoir 30 l Reservoir Smart2Pure 12 Pezzo
11307091 Feed Water Tank 30 l Tank GenPure, GenPure Pro, GenPure xCAD Pezzo
11397091 Storage Reservoir 100 L Level Display Pacific RO, Pacific TII Pezzo
11337091 Storage Reservoir 60 L Level Display Pacific RO, Pacific TII Pezzo
11387091 Storage Reservoir 100 L Level Display, Recirculation Pump Pacific TII Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Product Code. 13498709 Supplier Thermo Scientific™ Supplier No. 06.5001
  • ONLINE EXCLUSIVE PRICE
    187.00€
    179.00€ / pezzi 1
    Risparmia 8.00€
    4% Sconto
In stock
Add to basket
Edge
Add to basket
I serbatoi compatibili con i sistemi Barnstead™ GenPure™, Pacific™ o Smart2Pure™ Thermo Scientific™ consentono di conservare in sicurezza l'acqua purificata.
Safely store purified water from a compatible system with Thermo Scientific™ Water Purification Systems Storage Reservoirs. For use with Thermo Scientific™ Barnstead™ GenPure™, Pacific™ or Smart2Pure™ systems, these reservoirs are compatible with the following models: GenPure, GenPure Pro, GenPure xCAD, GenPure xCAD Plus, Pacific RO, Pacific TII and Smart2Pure.
Features:
  • Large opening offers easy and effective cleaning.
  • Optional safety filters include a CO2 absorber prevents conductivity increases from drawn-in CO2 and a sterile overflow to prevent microorganisms from entering and contaminating reservoir water.
  • Polyethylene reservoir is constructed with inert materials opaque to light.
  • Recirculation pump (optional) protects water from bacterial growth during standstill while maintaining a low conductivity value.
  • Wall mount is optional.
  • Conical bottom outlet allows for complete draining.
For use with GenPure, GenPure Pro, and GenPure xCAD purification systems only:
  • 06.5038
  • 06.5068
For use with Pacific RO water purification systems only:
  • 06.5034
  • 06.5064
  • 06.5084
For use with Pacific TII water purification systems only:
  • 06.5032
  • 06.5062
  • 06.5082
For use with Pacific RO & Pacific TII water purification systems only:
  • 06.5033
  • 06.5063
  • 06.5083
For use with Smart2Pure 12 water purification systems only:
  • 06.5040
  • 06.5070
TRUSTED_SUSTAINABILITY
Specifications
Capacità (sistema metrico) 30 to 100 L
Da utilizzare con (apparecchiatura) Smart2Pure
Tipo Sterile Overflow
Include N/A
Materiale Polyethylene