Learn More
ITS Thorsten Betzel™ WaterWorks™ Free Chlorine High Range
Product Code. 11722947
Change view
Click to view available options
Quantity:
50 Strips
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11722947 50 Strips Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11722947 Supplier ITS Thorsten Betzel™ Supplier No. 480022
  • ONLINE EXCLUSIVE PRICE
    21.14€
    17.77€ / pezzi 50
    Risparmia 3.37€
    15% Sconto
 Restricted Item
In stock
Add to basket
Edge
Add to basket
Use for determining whether or not you have the correct amount of sanitizer present. ITS Europe™ WaterWorks™ Free Chlorine High Range Strips combine a wide range of detection with patented Free Chlorine indicator.
  • Wide range of detection
  • Free Chlorine indicator
  • Designed to resist interference from monochloramines
  • Perfect for testing in a food processing environment
  • Range of detection, from 0-120 ppm (mg/L)
  • Perfect for determining whether or not you have the correct amount of sanitizer present.
  • Perfect to take directly out onto the shop floor; thus reducing the amount of rejections and misadjusted processes.
Specifications
Concentration Free Chlorine/1.0-5.0-10-20-40-80-120ppm
Packaging Bottle
Quantity 50 Strips