Learn More
Watson-Marlow Marprene Double Segment Manifold Tubing
Descrizione
- Made from autoclavable marprene
- For use with 400DM2 and 400DM3 pumpheads
- Can be used with any multichannel cartridges
- Adjacent cartridges may contain tubing of a different material or size
Specifica
Specifica
| Diametro (sistema metrico) interno | 0.25 mm |
| Materiale | Marprene |
| Da utilizzare con | 400DM2 and 400DM3 pumpheads |
| Dimensioni dei tubi | 0.25 mm |
| Product Type | Manifold Tubing |