missing translation for 'onlineSavingsMsg'
Learn More

WDR55 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marca:  Thermo Scientific PA561665

Dettagli aggiuntivi : Peso : 0.02000kg

Codice prodotto. 15779471

  • 510.00€ / 100µL
Spedizione stimata: 10-04-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Immunogen sequence: VSKAHGAPIN SLLLVDENVL ATGDDTGGIR LWDQRKEGPL MDMRQHEEYI ADMALDPAKK LLLTASGDGC LGIFNIKRRR FELLSEPQSG DLT Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 96%.

WDR55 Polyclonal antibody specifically detects WDR55 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).
Specifica

Specifica

WDR55
Polyclonal
Unconjugated
WDR55
WD repeat domain 55; WD repeat-containing protein 55; WDR55
Rabbit
Antigen affinity chromatography
RUO
54853
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin)
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9H6Y2
WDR55
Recombinant protein corresponding to Human WDR55
100 μL
Primary
Human
Antibody
IgG
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali

Research Use Only