Learn More
Welch™ DuoSeal™ Pump Oil
Product Code. 12835353 Shop All Welch Ilmvac Products
Change view
Click to view available options
Volume (Metric):
3.78 L
Unit Size:
3.7 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
12835353 3.78 L 3.7 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12835353 Supplier Welch™ Supplier No. U1407K15
  • ONLINE EXCLUSIVE PRICE
    108.00€
    106.50€ / 3.7L
    Risparmia 1.50€
    1% Sconto
View all promotions
 Restricted Item
Spedizione stimata: 21-12-2026
For use in DuoSeal High-Vacuum Pumps
For use in DuoSeal High-Vacuum Pumps. One gallon.Specially fractionate oil has properties enabling highest performance for DuoSeal pumps
  • Low vapor Pressure
  • High Viscosity
TRUSTED_SUSTAINABILITY
Specifications
Product Type Vaccum Pump Oil
For Use With (Application) Provides the lubrication that helps enable renowned DuoSeal™ pump durability and ensure the highest vacuum performance
For Use With (Equipment) DuoSeal™ Belt-driven pumps
Quantity 1 Ea.
Volume (English) 1 gal.
Volume (Metric) 3.78 L
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.