Learn More
Cytiva Whatman™ Adsorption 60Å Silica Gel TLC Plates
Codice prodotto. 11390804
Cambia vista
Click to view available options
Dimensioni (L x P):
20 x 20 cm
Dimensione della confezione:
pezzi 25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Dimensioni (L x P) unitSize
11390804 20 x 20 cm pezzi 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 11390804 Fornitore Cytiva N. del fornitore 4860820
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi
Provide quality separation of moderately to strongly polar compounds, including carbohydrates, antibiotics, proteins, amino acids, and phospholipids
  • 250μm layer thickness
  • 2.5 × 7.6cm size is a microscope slide plate
  • With organic binder and moderate layer hardness for convenient spot recovery with a sample recovery tube
  • Versions with Linear-K preadsorbent strip have a 3cm strip of inert material that acts as an inactive blotter or staging area for rapid application of samples along the edge of standard silica plate
TRUSTED_SUSTAINABILITY
Specifica
Assorbente Partisil K6 60A
Dimensioni (L x P) 20 x 20 cm