Learn More
Cytiva Whatman™ Anotop™ IC 25mm Syringe Filter
Codice prodotto. 11374734
Cambia vista
Click to view available options
Dimensione del poro:
0.2 μm
Dimensione della confezione:
pezzi 200
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Dimensione del poro unitSize
11374734 0.2 μm pezzi 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 11374734 Fornitore Cytiva N. del fornitore 68099244
Can be used with most organic solvents and aqueous materials and are suitable for sample volumes up to 100mL
The distinctive hexagonal housing is manufactured from pigment-free polypropylene to eliminate sample contamination. No wetting agents or adhesives are used in the manufacturing process.
Anotop Syringe Filters contain the proprietary alumina-based Anopore membrane. Glass microfiber prefilter versions are available for difficult-to-filter samples.
  • Each batch certified for ion chromatography (IC)
  • Enhanced consistency of analytical results
  • Extended column life
  • Certified and guaranteed low levels of anion leaching for improved results
  • Inlet: female Luer-Lok™
  • Outlet: male slip Luer
  • Use for HPLC aqueous/organic sample preparation, cold sterilization of growth media, solvent filtration for spectroanalysis, phage and virus filtration, liposome extrusion, removal of high molecular weight proteins or polymers, colloid removal from deionized water
TRUSTED_SUSTAINABILITY
Specifica
Materiale Polypropylene
Dimensione del poro 0.2 μm
Sterilità Non-sterile
Quantità 250/Pk.
Tipo IC Syringe Filter
Diametro (sistema metrico) 10 mm