Learn More
QIAGEN Foam Swabs, Sterile
Product Code. 11380844
Change view
Click to view available options
Quantity:
100 Pk.
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11380844 100 Pk. Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11380844 Supplier QIAGEN Supplier No. WB100032
  • ONLINE EXCLUSIVE PRICE
    255.00€
    122.97€ / pezzi 100
    Risparmia 132.03€
    51% Sconto
Spedizione stimata: 11-12-2026
Add to basket
Add to basket
For buccal and saliva samples.
  • Sterile foam swabs for easy collection of buccal cells and saliva and transfer to e.g. QIAcard FTA card formats.
  • The nonabrasive head of the foam tipped applicator is the same size as the sample area on QIAcard FTA Cards to facilitate sample application.
Tekniske data
Type Foam-tipped Applicator
For Use With (Application) For the collection of saliva and buccal cells
Quantity 100 Pk.
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.