Learn More
Cytiva™ Whatman™ Gel Blotting Paper Grade GB003
Product Code. 12415272
Change view
Click to view available options
Length (Metric):
140 mm
150 mm
Width (Metric):
100 mm
110 mm
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Width (Metric) Unit Size
12415272 150 mm 100 mm Pack of 100
10646294 140 mm 110 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12415272 Supplier Cytiva™ Supplier No. 10427805
  • ONLINE EXCLUSIVE PRICE
    45.05€
    42.45€ / pezzi 100
    Risparmia 2.60€
    5% Sconto
Spedizione stimata: 14-12-2026
A thick paper ideal for lysis/denaturation of colony or plaque lifts and Western blots
  • High absorbency
  • Pure cellulose
  • No contamination during transfers
  • Fo chromatographic and blotting techniques
TRUSTED_SUSTAINABILITY
Specifications
Length (Metric) 150 mm
Width (Metric) 100 mm
For Use With (Application) Suitable for (lysis/denaturation of colony or plaque lifts and Western blots)
Format Sheet
Material Cellulose
Dimensions (LxW) 10 x 15 cm
Grade GB003