missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Pyramid Folded Filter Papers
Pyramid (square) folded filter paper is easy to use and saves valuable time when undertaking repetitive or multiple analyses.
Marca: Cytiva 989710112
Dettagli aggiuntivi : Peso : 1.00000kg
Descrizione
- Easily fits into conical funnels
- Saves valuable time when undertaking repetitive or multiple analyses
- Provides the same high quality and consistency
Specifica
1000/Pk. | |
Filter Paper | |
Cellulose | |
180 μm | |
87 g/m2 |
Nominal Ash Content: 0.06% | |
1 | |
Folded | |
125 mm |