Learn More
Cytiva Whatman™ Soil Analysis Filter Papers
Whatman™ Grade 515 ½ Soil Analysis Filter Paper, Folder are pre-pleated low phosphate papers for the filtration of calcium lactate extracts from soil samples for the determination of phosphorus and potassium.
Marca:  Cytiva 10310645
Dettagli aggiuntivi : Peso : 0.17000kg
Codice prodotto. 11353005
  • 73.90€ / pezzi 100
Spedizione stimata: 18-04-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • Filter papers for use in soil analysis
  • For K and P determination according to Egnér, Riehm, and Lederle
  • Pre-pleated for use in conical filters with increased exposed surface area
Specifica
Specifica
76 g/m2
Pre-pleated
375 s
Grade 512 ½
160 μm
150 mm
Grade 512 ½ Application-Specific Filter, folded, 150 mm
Cellulose
For the filtration of calcium lactate extracts from soil samples for the determination of K and P according to Egnér, Riehm and Lederle.
100 Pack
Filter Paper