Learn More
DWK Life Sciences WHEATON™ Complete Cyanide Distillation Apparatus: Components
Product Code. 12491551 Shop All Wheaton Science Products Products
Change view
Click to view available options
Type:
Absorber Top
Cold Finger Condenser
Cold Finger Jacket
Distilling Flask
Inlet Tube
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
12491551 Distilling Flask Each
12481551 Cold Finger Jacket Each
12471551 Cold Finger Condenser Each
12461551 Absorber Top Each
12401561 Inlet Tube Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 12491551 Supplier DWK Life Sciences Supplier No. 377165
  • ONLINE EXCLUSIVE PRICE
    157.00€
    148.35€ / pezzi 1
    Risparmia 8.65€
    5% Sconto
Spedizione stimata: 07-06-2027
WHEATON™ Complete Cyanide Distillation Apparatus: Components are replacement parts designed for use with the WHEATON™ Complete Cyanide Distillation Apparatus. Suitable for quantitative cyanide analysis of industrial wastewaters, saline, and reagent grade water per ASTM D2036.
  • Replacement component parts for the Wheaton Complete Cyanide Distillation Apparatus (Cat. No. 377160)
  • Designed for quantitative analysis of cyanides in industrial wastewaters, saline, and reagent grade water
  • Supports testing per ASTM D2036 Standard Test Method for Cyanides in Water
  • Components include absorber tube, absorber top, cold finger, cold finger jacket, distilling flask, and inlet tube
  • Standard taper ground glass joints (19/38 and 29/42) ensure secure, leak-resistant connections
  • Manufactured from borosilicate glass for chemical resistance and durability
Compliance
For use according to ASTM D 2036
Specifications
Volume (Metric) 1000 mL
For Use With (Equipment) Cyanide distillation kit
Type Distilling Flask
Volume (English) 33.81 oz.