Learn More
DWK Life Sciences WHEATON™ Peristaltic Pump Tubing: Hose Barb/Luer Adapter
Product Code. 12471531 Shop All Wheaton Science Products Products
Change view
Click to view available options
Diameter (Metric) Inside:
2 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inside Unit Size
12471531 2 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12471531 Supplier DWK Life Sciences Supplier No. 374322
  • ONLINE EXCLUSIVE PRICE
    41.50€
    31.06€ / pezzi 1
    Risparmia 10.44€
    25% Sconto
Spedizione stimata: 01-03-2027
WHEATON™ Peristaltic Pump Tubing: Hose Barb/Luer Adapter is designed for use with WHEATON™ Omnispense™ Elite and Unispense™ PRO peristaltic pumps, enabling reliable dispensing and manifold attachment.
  • Compatible with Wheaton Omnispense™ Elite and Unispense™ PRO peristaltic pumps
  • Designed for dispensing pump applications
  • Enables attachment of manifold assemblies
Specifications
Diameter (English) Inside 7/100 in.
Diameter (Metric) Inside 2 mm
For Use With (Application) Dispensing Pump
For Use With Dispensing Pump
Overall Length (English) 0.12 in.
Overall Length (Metric) 3 mm
Type Hose Barb/Luer Adapter
Length (Metric) 3 mm