Description
- Breathable back panel to help improve air ventilation and reduce heat build-up.
- Knit cuffs provide a soft wrist closure for increased comfort compared to elastic closures.
- Elastic waist and ankles for convenience and freedom of movement.
- Two-way zipper with sealable storm flap for convenience and to help provide additional protection.
Specifications
Specifications
| Product Type | General Purpose Coveralls |
| Size | Medium |
| Sesso | Unisex |
| Colore | Blue, White |
| Monouso | Single-use |
| Stile vita | Elastic |
| Stile polso | Knit |
| Stile collo | Hooded |
| Sterilità | Non-sterile |
| Modello | 4535W |