Learn More
MP Biomedicals™ Whole Serum
Product Code. 11474305 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
2 mL
Unit Size:
2mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11474305 2 mL 2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11474305 Supplier MP Biomedicals™ Supplier No. 0855979
  • ONLINE EXCLUSIVE PRICE
    353.00€
    336.00€ / 2mL
    Risparmia 17.00€
    4% Sconto
 Restricted Item
Spedizione stimata: 21-12-2026
Suitable for use as a serum protein control in immunoelectrophoresis, Ouchterlony immunodiffusion, and other immunoprecipitation techniques.
Specifications
Buffer 0.02M sodium phosphate, 0.14M sodium chloride, pH 7.3, filtered through a 0.22 μmfilter, vialed and lyophilized.
Description Whole Human Serum
Quantity 2 mL