Learn More
Cole-Parmer™ Wildco™ Kemmerer Water Sampler
Codice prodotto. 11774499 Sfoglia Tutto Cole-Parmer Prodotti
Cambia vista
Click to view available options
Capacità (sistema metrico):
400 mL
1200 mL
Dimensione della confezione:
Pezzo
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Capacità (sistema metrico) unitSize
11774499 400 mL Pezzo
11784499 1200 mL Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 11774499 Fornitore Cole-Parmer™ N. del fornitore 0548500
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Sample at specific depths between 3 and 600 feet.
  • WildcoKemmerer Water Sampler
  • The messenger activates the unique trip heads that ensure closure in fast flowing streams or turbulent waters, regardless of line angle
  • All models include a plastic carrying case; order messenger and line separately.
  • The 304 SS models have urethane end seals (do not use when mercury concentrations exceed 1 ppm).
Specifica
Product Type Kemmerer Water Sampler
Materiale Stainless Steel
Descrizione 3 to 600 ft depth
Capacità (sistema metrico) 400 mL