Learn More
Bel-Art™ SP Scienceware™ Wilmad™ Disposable NMR Tube Caps, 4.0mm Dia., Blue
Different colors help track samples based on various reference solvents.
Marca:  Bel-Art™ 66004-1001
Dettagli aggiuntivi : Peso : 0.21540kg
Codice prodotto. 15338708
  • 64.65€ / 1000 confezioni
Per tempo di consegna, consultarci allo 02 950 59 478
Aggiungi al carrello
Descrizione
Descrizione
Alert: When using Chloroform or Acetone as a reference solution, please avoid direct contact between the sample and the disposable cap, and avoid long term storage over 2 hours. If these conditions cannot be avoided, please use PTFE tube caps.
Specifica
Specifica
4.0mm
Polyethylene
NMR Spectroscopy
Disposable
Model: Wilmad™ 521-G-1000
Blue
1000 per pack
Videos