Learn More
Heinz Herenz Wooden Applicator With Cellulose Bud Standard Tip
Descrizione
We offer a wide range of sterile and nonsterile swabs:
- Wood, PS, PP, aluminium or paper applicator
- Each in various lengths available
- With cotton, viscose, polyester or foam head
- Diameter of the heads: small, medium or large
- This guarantees that the right instrument is available for every special application.
- For example, the cotton-tipped swabs are recommended for use by ENT specialists.
- Our sterile cotton buds are all individually wrapped.
Please note in the medical field:
- Only suitable for single use
- Use by medical personnel only
Specifica
Specifica
| Autoclavabile | Not Autoclavable |
| Materiale maniglia | Wood |
| Product Type | Applicator Stick |
| Tip Material | Wood |
| Tipo di punta | Standard |