Learn More
Wrench, universal, suited for Acros drums, , Thermo Scientific™
Product Code. 17790689 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
For Use With (Equipment):
Acros Drums
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
17790689 Acros Drums Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17790689 Supplier Thermo Scientific Acros Supplier No. 473470010
 Restricted Item
In stock
Specifications
Color Silver
For Use With (Application) Open Sealed Caps and Drum Seals
For Use With (Equipment) Acros Drums
Material Mild Steel
Dimensions 256 x 100 x 45 mm
Length (Metric) 256 mm
Width (Metric) 100 mm
Height (Metric) 45 mm
Weight (Metric) 1 Kg