Learn More
ABEBA™ X-Light Black Occupational Shoes O2 ESD
Product Code. 17191525
Change view
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17191525 40 1 pair
17141525 35 1 pair
17151525 36 1 pair
17141535 45 1 pair
17151535 46 1 pair
17161535 47 1 pair
17171535 48 1 pair
17161525 37 1 pair
17171525 38 1 pair
17181525 39 1 pair
17101535 41 1 pair
17111535 42 1 pair
17121535 43 1 pair
17131535 44 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 17191525 Supplier ABEBA™ Supplier No. 7131127/40
Spedizione stimata: 22-12-2026
Black X-light occupational shoes with silver ion lining to reduce odor, replaceable active comfort insole, steel toe cap, PU slip resistant outsole
  • Lightweight, comfortable black occupational shoes
  • Comply to EN ISO 20347: 2012, O2 SRC
  • ESD dissipative according EN 61340 class climatic 1
  • Slip resistant Mono PU outsole SRC
  • Black microfiber
  • Silver ions lining
  • Replaceable active comfort insole
Specifications
Color Black
Material Microfiber
Size 40
Certifications/Compliance (Europe) Comply to EN 20347: 2012 O2 SRC, ESD dissipative according EN 61340
Gender Unisex
Ankle Style Low-cut
Waterproof Water Resistant
Product Type O2 SRC ESD