Learn More
Honeywell Safety Products™ XC Rx Insert Safety Glasses
Descrizione
- Rx Insert fits securely behind the spectacle lens.
- XC is certified for impact against EN 166, FT: excellent impact protection (particles at 45m/s) in extreme hot or cold working conditions.
- XC offers high level of UV filtration protection: EN 166, 2C-1.2.
- Less expensive than an Rx safety frame.
- Reduces number of eyewear styles per plant.
- Designed to fit a wide variety of prescription lenses.
- XC lens protects Rx lens, greatly reducing high cost of replacing Rx lens.
- Adjustability and soft material not available on most Rx frames.
- More attractive and less bulky than current over-the-glasses eyewear.
Specifica
Specifica
| Certificazione/Conformità | EC Category PPE 2, EC Certificate Number EC1144, EC Attestation Number EC1144 |
| Product Type | Safety Glasses |
| N. per confezione | 1 ciasc. |
| Materiale del telaio | Metal Alloy (Nickel-free plating) |