Learn More
Trajan Scientific and Medical™ XCHANGE™ Syringes for use with MEPS BINS
Codice prodotto. 15289263
Cambia vista
Click to view available options
Volume (sistema metrico):
100 μL
250 μL
Dimensione della confezione:
Pezzo
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Volume (sistema metrico) Dimensione della confezione
15289263 100 μL Pezzo
15219273 250 μL Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 15289263 Fornitore Trajan Scientific and Medical™ N. del fornitore 2928500
Spedizione stimata: 14-12-2026
With PTFE-tipped plunger
  • High quality solutions for liquid handling
  • For use on CTC PAL-xt
Accurate and reproducible injectionsXCHANGE Syringes specifically designed for MEPS Applications
Specifica
Materiale PTFE-tipped (Plunger)
Diametro (sistema metrico) esterno 6.7 mm syringe barrel
Volume (sistema metrico) 100 μL