missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XRCC3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 18494-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This gene encodes a member of the RecA/Rad51-related protein family that participates in homologous recombination to maintain chromosome stability and repair DNA damage. This gene functionally complements Chinese hamster irs1SF, a repair-deficient mutant that exhibits hypersensitivity to a number of different DNA-damaging agents and is chromosomally unstable. A rare microsatellite polymorphism in this gene is associated with cancer in patients of varying radiosensitivity. Alternatively spliced transcript variants encoding the same protein have been identified.Specifica
XRCC3 | |
Polyclonal | |
Unconjugated | |
XRCC3 | |
DNA repair protein XRCC3, XRCC3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
102551085, 74335, 7517 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot, Immunofluorescence | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O43542, Q9CXE6 | |
Xrcc3 | |
XRCC3 Fusion Protein Ag13317 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |