Learn More
Chemsplash Xtreme 50 SMS Flame Retardant A/S C/All Type 5/6 White
GREENER_CHOICE
Codice prodotto. 30058440
Cambia vista
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Dimensione della confezione:
pezzi 25
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Size unitSize
30058440 3X-Large pezzi 25
30058304 Small pezzi 25
30058404 Medium pezzi 25
30058367 Large pezzi 25
30058194 X-Large pezzi 25
30058434 2X-Large pezzi 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Codice prodotto. 30058440 Fornitore Chemsplash N. del fornitore 2513.400.009
Aggiungi al carrello
Aggiungi al carrello
The Xtreme SMS 50 Flame Retardant Disposable Coverall is made from a breathable SMS fabric which has been chemically treated to offer limited flame retardant protection to EN14116 index 1.
Chemsplash Xtreme SMS 50 Flame Retardant Disposable Coverall
  • Material: 50GSM Breathable SMS Fabric
  • Features:
    • 3-Piece Elasticated Hood, Wrists, Back, and Ankles
    • Two-Way Zip with Adhesive Zip Flap
    • Anti-Static
    • Flame Retardant to EN14116 Index 1
    • Flame Retardancy Includes All Material Assemblies
    • Silicone, Latex Free and PFAS Free
    • Comes Without the Chest Label
  • Conforms To: CAT III Type 5/6
The Chemsplash Xtreme SMS 50 Flame Retardant Disposable Coverall is made from breathable SMS fabric treated to provide limited flame retardant protection to EN14116 Index 1. This includes all material assemblies, meeting the 2016/425 regulation. This suit features an elasticated hood, wrists, ankles, and a two-way zip with an adhesive flap cover. Its robust and comfortable design provides excellent chemical barrier protection. It is suitable to be worn over a primary EN531 flame retardant cotton-based coverall without compromising the flame-resistant properties of the EN531 garment.
Note: Disposable FR garments that meet EN14116 or its predecessor standard EN533:1997 should never be used as primary (sole) flame retardant protection.
Specifica
Product Type Hooded Flame-resistant Coveralls
Size 3X-Large
Sesso Unisex
Materiale FR Anti-static SMS Fabric
Colore White
Monouso Single-use
Stile vita Elastic
Stile della caviglia Elastic
Stile polso Elastic
Stile collo Hooded
Tipo di cappa 3-panel
Tipo di chiusura Ties
Tipo di giuntura Stitched
Sterilità Non-sterile
Resistenza Particle, Flame
Senza lattice Yes
N. di tasche None
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Select an issue
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.