missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YARS2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA550344
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
The antibody detects endogenous levels of total YARS2 protein.
YARS2 Polyclonal antibody specifically detects YARS2 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifica
YARS2 | |
Polyclonal | |
Unconjugated | |
YARS2 | |
2210023C10Rik; CGI-04; EC 6.1.1.1; fi41d10; LOC556702 protein; LOW QUALITY PROTEIN: tyrosine--tRNA ligase, mitochondrial; MLASA2; MT-TYRRS; RGD1311696; tyrosine tRNA ligase 2, mitochondrial; tyrosine--tRNA ligase, mitochondrial; tyrosyl trna synthetase; Tyrosyl-tRNA synthetase; tyrosyl-tRNA synthetase 2; tyrosyl-tRNA synthetase 2 (mitochondrial); tyrosyl-tRNA synthetase 2, mitochondrial; tyrosyl-tRNA synthetase mitochondrial-like protein; tyrosyl-tRNA synthetase, mitochondrial; tyrRS; wu:fi41d10; Yars2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51067, 70120 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q8BYL4, Q9Y2Z4 | |
YARS2 | |
Fusion protein corresponding to residues near the C terminal of human tyrosyl-tRNA synthetase 2, mitochondrial | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |