Learn More
Thermo Scientific Chemicals Yeast Extract
Product Code. 15483079
Change view
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15483079 500 g 500g
15493079 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15483079 Supplier Thermo Scientific Chemicals Supplier No. J60287.36
 Restricted Item
In stock
Add to basket
Edge
Used as food additive, flavor enhancer in processed foods of all kinds and as nutrient for bacterial culture media. It is used to create savoury flavors and umami taste sensations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
TRUSTED_SUSTAINABILITY
Specifications
Form Powder
Packaging Type Plastic Bottle
Quantity 500 g