Learn More
3M™ Yellow Headgear
Product Code. 15150229 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
Faceshield 5F-11 and 3M Peltor Optime I Earmuff
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15150229 Faceshield 5F-11 and 3M Peltor Optime I Earmuff Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15150229 Supplier 3M™ Supplier No. 7000038216
  • ONLINE EXCLUSIVE PRICE
    380.00€
    300.31€ / pezzi 10
    Risparmia 79.69€
    20% Sconto
Spedizione stimata: 05-02-2027
Specifications
Color Yellow
For Use With (Equipment) Faceshield 5F-11 and 3M Peltor Optime I Earmuff
Product Type Headgear
Quantity 10/Pk.