Learn More
Checkers Yellow Jacket™ 3-Channel AMS Cable Protector Kit
Product Code. 17762428
Change view
Click to view available options
Color:
Black, Yellow
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17762428 Black, Yellow Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17762428 Supplier Checkers Supplier No. PYJS0003YB
  • ONLINE EXCLUSIVE PRICE
    3198.20€
    2598.86€ / pezzi 1
    Risparmia 599.34€
    18% Sconto
 Restricted Item
Spedizione stimata: 26-11-2026
Yellow Jacket™ 3-channel AMS cable protector kit features a gradual slope and slip-resistant surface to ensure a safer crossing point for carts, forklifts, vehicles with small wheels and heavy trucks.
  • Yellow Jacket™ 3-channel AMS cable protector kit features a gradual slope and slip-resistant surface to ensure a safer crossing point for carts, forklifts, vehicles with small wheels and heavy trucks
  • In addition, each Accessibility Ramp system is manufactured to meet ADA and DDA compliant specifications
Specifications
Length (English) 6.5 ft.
Length (Metric) 198 cm
Width (Metric) 181 cm
Color Black, Yellow
Material Polyethylene
Type Cable Protector Kit