Learn More
Checkers Yellow Jacket™ AMS™ 5 Channel Male Ramps
Product Code. 17792428
Change view
Click to view available options
Color:
Black
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17792428 Black Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17792428 Supplier Checkers Supplier No. PYJS0006BL
  • ONLINE EXCLUSIVE PRICE
    145.37€
    118.44€ / pezzi 1
    Risparmia 26.93€
    18% Sconto
Spedizione stimata: 26-11-2026
Yellow Jacket™ 5-channel AMS ramps can be combined with the Yellow Jacket™ 5-channel AMS center section to create a system that is fully expandable to hold an unlimited number of cables.
  • Yellow Jacket™ 5-channel AMS ramps can be combined with the Yellow Jacket™ 5-channel AMS center section to create a system that is fully expandable to hold an unlimited number of cables
  • By detaching the ramps, you can also quickly add cable runs and ADA-compatible side ramps to existing systems
  • The Yellow Jacket™ AMS™ readily handles heavy truck and bus traffic while providing easy access for pedestrians, carts, and wheelchairs
  • Hinged lids make for easy loading of cables and hoses
Specifications
Dimensions (L x W x H) 51 x 91 x 5 cm
Color Black
Material Polyethylene
Type 5 Channel Ramps