Learn More
Yoda 1, TRC
Product Code. 30453695
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30453695 5 mg 5mg
30456560 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30453695 Supplier Toronto Research Chemicals Supplier No. TRCY1007855MG
 Restricted Item
Spedizione stimata: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 448947-81-7
Molecular Formula C13H8Cl2N4S2
Molecular Weight (g/mol) 355.27
Synonym 2-[5-[[(2,6-Dichlorophenyl)methyl]thio]-1,3,4-thiadiazol-2-yl]pyrazine, [5-[[(2,6-Dichlorophenyl)methyl]thio]-1,3,4-thiadiazol-2-yl]-pyrazine
IUPAC Name 2-[(2,6-dichlorophenyl)methylsulfanyl]-5-pyrazin-2-yl-1,3,4-thiadiazole
SMILES Clc1cccc(Cl)c1CSc2nnc(s2)c3cnccn3
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C13H8Cl2N4S2/c14-9-2-1-3-10(15)8(9)7-20-13-19-18-12(21-13)11-6-16-4-5-17-11/h1-6H,7H2
Formula Weight 353.9567
Chemical Name or Material Yoda 1