Learn More
YSI™ Pro Plus Quatro Field Cable
Codice prodotto. 11968089
Cambia vista
Click to view available options
Da utilizzare con (apparecchiatura):
Professional Plus instrument
Dimensione della confezione:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Da utilizzare con (apparecchiatura) Dimensione della confezione
11968089 Professional Plus instrument Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 11968089 Fornitore YSI™ N. del fornitore 60579030
Quantità Prezzo
1 2759.10€ / pezzi 1
3 2188.52€ / pezzi 1
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi
Includes
Conductivity/temperature sensor
For use with Professional Plus instrument.
  • 4-port cable
  • True field cable with MS (military spec) connectors
  • Rugged, weighted probe guard
  • Measure conductivity/temperature, dissolved oxygen and any two ISEs among pH, ORP, ammonium, nitrate or chloride
  • Cable Length: 30m
Warranty
Two years
TRUSTED_SUSTAINABILITY
Specifica
Da utilizzare con (apparecchiatura) Professional Plus instrument