Learn More
YSI™ Probe Service Kits for YSI Dissolved Oxygen Probes
Product Code. 11978876
Change view
Click to view available options
Type:
High-sensitivity Membrane Kit
Standard Membrane Kit
Quantity:
10 x 150 Membranes
30 Membranes/Pk.
Unit Size:
Each
Pack of 10
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Quantity Unit Size
11978876 Standard Membrane Kit 30 Membranes/Pk. Pack of 10
11969176 High-sensitivity Membrane Kit 10 x 150 Membranes Pack of 10
11988876 High-sensitivity Membrane Kit 30 Membranes/Pk. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11978876 Supplier YSI™ Supplier No. 098094
 Restricted Item
Spedizione stimata: 07-12-2026
For replenishing oxygen sensor on YSI Dissolved Oxygen Probes
  • Membrane Kits include 150 standard, 1mil PTFE membranes or 150 high-sensitivity, 0.5mil PTFE membranes, in 10 packs of 50
  • Membrane/KCI Kits include 30 standard or high-sensitivity membranes, 1 oz. (30mL) dropper bottle with KCI crystals for adding distilled water and a probe contour tool
  • For use with probes 13-299-43 (Model 5739), 13-298-80 (Model 5750), 15-176-50 (Model 5718), 13-298-16 (Model 5300 Biological Oxygen Monitor), and 13-298-56, -57, or -62 (Model 55 Handheld Dissolved Oxygen Meters)
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) 5718, 5739, 5750, 5300 and Rapid Pulse Probes
Type Standard Membrane Kit
Includes Bottle of electrolyte, 2 O-rings, 30 1mL FEP Fluoropolymer Film Membranes
Probe Type DO
Quantity 30 Membranes/Pk.