missing translation for 'onlineSavingsMsg'
Learn More

Ytterbium foil, 0.64mm (0.025in) thick, 99.9% (REO), Thermo Scientific Chemicals

Catalog Number p-7048237
Click to view available options
Quantità:
25 x 25 mm
50 x 50 mm
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7440-64-4
Yb
173.05
MFCD00011286
NAWDYIZEMPQZHO-UHFFFAOYSA-N
unii-mnq4o4wsi1, hydride, mnq4o4wsi1, foil, 3n, yterbio, ingot, powder, ii ion, chips, 3n, ingot, 3n
23992
CHEBI:33381
ytterbium
[Yb]
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7440-64-4
Yb
173.05
MFCD00011286
NAWDYIZEMPQZHO-UHFFFAOYSA-N
unii-mnq4o4wsi1, hydride, mnq4o4wsi1, foil, 3n, yterbio, ingot, powder, ii ion, chips, 3n, ingot, 3n
23992
CHEBI:33381
ytterbium
[Yb]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY
Materiale o nome chimico Ytterbium foil
Nota nome 0.64mm (0.025 in.) thick
Note percentuale saggio (REO)
Quantità 25 x 25 mm
Percent Purity 99.9%
Imballaggio Mylar under Argon
Peso ≈2.8g/25x25mm
Forma Foil

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'