Learn More
(Z)-Tritriacont-24-en-16-one, TRC
Product Code. 30454910
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30454910 5 mg 5mg
30450866 25 mg 25mg
30451884 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30454910 Supplier Toronto Research Chemicals Supplier No. TRCT7411405MG
 Restricted Item
Spedizione stimata: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 71177-09-8
Molecular Formula C33H64O
Molecular Weight (g/mol) 476.86
IUPAC Name (Z)-tritriacont-24-en-16-one
SMILES CCCCCCCCCCCCCCCC(=O)CCCCCCC\C=C/CCCCCCCC
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C33H64O/c1-3-5-7-9-11-13-15-17-18-20-22-24-26-28-30-32-33(34)31-29-27-25-23-21-19-16-14-12-10-8-6-4-2/h17-18H,3-16,19-32H2,1-2H3/b18-17-
Formula Weight 476.4957
Chemical Name or Material (Z)-Tritriacont-24-en-16-one