Learn More
ZAN Monoclonal Antibody (2B6), Invitrogen™
Mouse Monoclonal Antibody
Marca:  Invitrogen MA524422
Dettagli aggiuntivi : Peso : 0.01000kg
Codice prodotto. 15602255
  • 472.00€ / 100µg
Spedizione stimata: 17-05-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
Peptide Sequence: KEKPPDQKLV VRSSRDNYVL TQCDFEDDAK PLCDWSQVSA DDEDWVRASG PSPTGSTGAP GGYPNGEGSY LHMESNSFHR GGVARLLSPD LWEQG
ZAN Monoclonal antibody specifically detects ZAN in Human samples. It is validated for ELISA, Western Blot.
Specifica
Specifica
ZAN
Monoclonal
0.5 mg/mL
PBS with no preservative; pH 7.4
Q9Y493
ZAN
ZAN (NP_003377.1, 19 a.a. ∽ 113 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Primary
Human
Antibody
IgG1 κ
ELISA, Western Blot
2B6
Unconjugated
ZAN
ZAN; Zonadhesin; zonadhesin (gene/pseudogene)
Mouse
Protein A
RUO
7455
-20° C, Avoid Freeze/Thaw Cycles